Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
MA1-10857 detects IL-1 Receptor from human samples.
MA1-10857 has been successfully used in ELISA, radioimmune assays, and immunohistochemistry applications.
The MA1-10857 immunogen is recombinant extracellular domain of the type I human IL-1 receptor.
The IL-1 system includes two agonists (IL-1 and IL-1), converting enzymes, antagonists, two receptors (IL-1 RI and IL-1 RII) and the IL-1 receptor accessory protein. Interleukin-1 signal is transduced through the type I receptor.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: antigen CD121a; CD121 alpha; CD121 antigen-like family member A; CD121a; il-1 receptor; il-1r; IL-1R-1; IL-1R-alpha; IL-1RT-1; IL-1RT1; il1 receptor; interleukin 1 receptor alpha, type I; interleukin 1 receptor, type I; interleukin-1 receptor; Interleukin-1 receptor alpha; Interleukin-1 receptor type 1; Interleukin-1 receptor type I; p80
Gene Aliases: CD121A; D2S1473; IL-1R-alpha; IL1R; IL1R1; IL1RA; IL1RT1; P80
UniProt ID: (Human) P14778
Entrez Gene ID: (Human) 3554
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support