Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: AARSCRFRG RHYKREFRLE GEPVALRCPQ VPYWLWASVS PRINLTWHKN DSARTVPGEE ETRMWAQDGA LWLLPALQED SGTYVCTTRN ASYCDKMSIE LRVFENTDAF LPFISYPQIL TLSTSGVLVC PDLSEFTRDK TDVKIQWYKD SLLLDKDNEK FLSVRGTTHL LVHDVALEDA GYYRCVLTFA HEGQQYNITR SIELRIKKKK EETIPVIISP LKTISASLGS RLTIPCKVFL GTGTPLTTML WWTANDTHIE SAYPGGRVTE GPRQEYSENN ENYIEVPLIF DPVTREDLHM DFKCVVHNTL SFQTLRTTVK EASST (24-347 aa encoded by B C039031)
The protein encoded by this gene is a cytokine receptor that belongs to the interleukin 1 receptor family. This protein binds interleukin alpha (IL1A), interleukin beta (IL1B), and interleukin 1 receptor, type I(IL1R1/IL1RA), and acts as a decoy receptor that inhibits the activity of its ligands. Interleukin 4 (IL4) is reported to antagonize the activity of interleukin 1 by inducing the expression and release of this cytokine. This gene and three other genes form a cytokine receptor gene cluster on chromosome 2q12. Two alternatively spliced transcript variants encoding the same protein have been reported.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: antigen CDw121b; CD121 antigen-like family member B; CD121b; CDw121b; IL-1 receptor beta chain; IL-1 type II receptor; IL-1R-2; IL-1R-beta; IL-1RT-2; IL-1RT2; ILR2; interleukin 1 receptor type II variant 3; interleukin 1 receptor, type II; Interleukin-1 receptor beta; Interleukin-1 receptor type 2; Interleukin-1 receptor type II; type II interleukin-1 receptor, beta
Gene Aliases: CD121b; CDw121b; IL-1R-2; Il-1r2; IL-1RT-2; IL-1RT2; Il1r-2; IL1R2; IL1R2c; IL1RB
UniProt ID: (Human) P27930, (Mouse) P27931
Entrez Gene ID: (Human) 7850, (Mouse) 16178
Molecular Function:
transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support