Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MTLLWCVVSL YFYGILQSDA SERCDDWGLD TMRQIQVFED EPARIKCPLF EHFLKFNYST AHSAGLTLIW YWTRQDRDLE EPINFRLPEN RISKEKDVLW FRPTLLNDTG NYTCMLRNTT YCSKVAFPLE VVQKDSCFNS PMKLPVHKLY IEYGIQRITC PNVDGYFPSS VKPTITWYMG CYKIQNFNNV IPEGMNLSFL IALISNNGNY TCVVTYPENG RTFHLTRTPT VKVVGSPKNA VPPVIHSPND HVVYEKEPGE ELLIPCTVYF SFLMDSRNEV WWTIDGKKPD DITIDVTINE SISHSRTEDE TRTQILSIKK VTSEDLKRSY VCHARSAKGE VAKAAKVKQK GNRCGQ
This gene encodes a component of the interleukin 1 receptor complex, which initiates signaling events that result in the activation of interleukin 1-responsive genes. Alternative splicing of this gene results in membrane-bound and soluble isoforms differing in their C-terminus. The ratio of soluble to membrane-bound forms increases during acute-phase induction or stress.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hide; IL 1R 3; IL 1R3; IL 1RAcP; IL 1RAP; IL-1 receptor accessory protein; IL-1R-3; IL-1R3; IL1RAcP; ILN; Interleukin; Interleukin-1 receptor 3; Interleukin-1 receptor accessory protein; interleukin-1 receptor accessory protein beta
Gene Aliases: C3orf13; IL-1RAcP; IL1R3; IL1RAP
UniProt ID: (Human) Q9NPH3
Entrez Gene ID: (Human) 3556
Molecular Function: transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support