Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%).
This antibody has specificity for Human IL-1R9/IL1RAPL2.
The protein encoded by this gene is a member of the interleukin 1 receptor family. This protein is similar to the interleukin 1 accessory proteins, and is most closely related to interleukin 1 receptor accessory protein-like 1 (IL1RAPL1). This gene and IL1RAPL1 are located at a region on chromosome X that is associated with X-linked non-syndromic mental retardation. [provided by RefSeq, Jul 2008].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: IL-1 receptor accessory protein-like 2; IL-1R-9; IL1RAPL-2-related protein; interleukin 1 receptor 9; interleukin 1 receptor accessory protein-like 2; Interleukin-1 receptor 9; Three immunoglobulin domain-containing IL-1 receptor-related 1; TIGIRR-1; X-linked interleukin-1 receptor accessory protein-like 2
Gene Aliases: IL-1R9; IL1R9; IL1RAPL-2; IL1RAPL2; TIGIRR-1
UniProt ID: (Human) Q9NP60
Entrez Gene ID: (Human) 26280
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support