Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MKHLLMWSP VIAPGETVYY SVEYQGEYES LYTSHIWIPS SWCSLTEGPE CDVTDDITAT VPYNLRVRAT LGSQTSAWSI LKHPFNRNST ILTRPGMEIT KDGFHLVIEL EDLGPQFEFL VAYWRREPGA EERPFPWYWP CLPLLASC (48-178 aa encoded by BC033292 )
IL20RB and IL20RA (MIM 605620) form a heterodimeric receptor for interleukin-20 (IL20; MIM 605619) (Blumberg et al., 2001 [PubMed 11163236]).[supplied by OMIM, Feb 2009].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Fibronectin type III domain containing 6; FNDC6; IL-20 receptor subunit beta; IL-20R-beta; IL-20R2; IL-20RB; interleukin 20 receptor beta subunit; interleukin-20 receptor II; Interleukin-20 receptor subunit beta
Gene Aliases: DIRS1; FNDC6; IL-20R2; IL20RB; UNQ557/PRO1114
UniProt ID: (Human) Q6UXL0
Entrez Gene ID: (Human) 53833
Molecular Function:
transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support