Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: GVSMPRGWAA PLLLLLLQGG WGCPDLVCYT DYLQTVICIL EMWNLHPSTL TLTWQDQYEE LKDEATSCSL HRSAHNATHA TYTCHMDVFH FMADDIFSVN ITDQSGNYSQ ECGSFLLAES IKPAPPFNVT VTFSGQYNIS WRSDYEDPAF YMLKGKLQYE LQYRNRGDPW AVSPRRKLIS VDSRSVSLLP LEFRKDSSYE LQVRAGPMPG SSYQGTWSEW S; Positive Samples: U-87MG, Mouse spleen, Mouse thymus; Cellular Location: Membrane, Single-pass type I membrane protein
The protein encoded by this gene is a cytokine receptor for interleukin 21 (IL21). It belongs to the type I cytokine receptors, and has been shown to form a heterodimeric receptor complex with the common gamma-chain, a receptor subunit also shared by the receptors for interleukin 2 (IL2) and interleukin 5 (IL5). This receptor transduces the growth promoting signal of IL21, and is important for the proliferation and differentiation of T cells, B cells, and natural killer (NK) cells. The ligand binding of this receptor leads to the activation of multiple downstream signaling molecules, including JAK1, JAK3, STAT1, and STAT3. Knockout studies of a similar gene in mouse suggest a role for this gene in regulating immunoglobulin production. Three alternatively spliced transcript variants encoding the same protein have been described.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD360; IL-21 receptor; IL-21R; Interleukin-21 receptor; LR-beta; Lymphocyte receptor beta; MGC10967; Novel cytokine receptor 8; Novel interleukin receptor; NR8
Gene Aliases: CD360; IL21R; NILR; UNQ3121/PRO10273
UniProt ID: (Human) Q9HBE5, (Mouse) Q9JHX3
Entrez Gene ID: (Human) 50615, (Mouse) 60504
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support