Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MRTLLTILTV GSLAAHAPED PSDLLQHVKF QSSNFENILT WDSGPEGTPD TVYSIEYKTY GERDWVAKKG CQRITRKSCN LTVETGNLTE LYYARVTAVS AGGRSATKMT DRFSSLQHTT LKPPDVTCIS KVRSIQMIVH PTPTPIRAGD GHRLTLEDIF HDLFYHLELQ VNRTYQMHLG GKQREYEFFG LTPDTEFLGT IMICVPTWAK ESAPYMCRVK TLPDRTWTYS FSGAFLFSMG FLVAVLCYLS YRYVTKPPAP PNSLNVQRVL TFQPLRFIQE HVLIPVFDLS GPSSLAQPVQ YSQIRVSGPR EPAGAPQRHS LSEITYLGQP DISILQPSNV PPPQILSPLS YAPNAAPEVG PPSYAPQVTP EAQFPFYAPQ AISKVQPSSY APQATPDSWP PSYGVCMEGS GKDSPTGTLS SPKHLRPKGQ LQKEPPAGSC MLGGLSLQEV TSLAMEESQE AKSLHQPLGI CTDRTSDPNV LHSGEEGTPQ YLKGQLPLLS SVQIEGHPMS LPLQPPSRPC SPSDQGPSPW GLLESLVCPK DEAKSPAPET SDLEQPTELD SLFRGLALTV QWES
A novel cytokine, designated IL-TIF for IL-10 related T cell-derived inducible factor and IL-22, was recently identified. The receptor for IL-22 (IL-22R, also termed CRF2-9 and IL-TIF-R1 chain) is a new member of the class II cytokine receptor family. IL-22R forms a complex with IL-10 receptor beta chain and mediates IL-22 signaling. IL-22 and its receptor activate JAK-STAT signaling pathway. IL22R is expressed in normal liver and kidney and their cell lines HepG2 and TK-10. A soluble form of IL-22 receptor, also termed IL-22 binding protein (IL-22BP) and IL-22 receptor-alpha 2 (IL-22RA2), was identified very recently. IL-22BP prevents binding of IL-22 to the functional cell surface IL-22R complex and neutralizes IL-22 activity. LPS induces IL-22 expression, which indicates the role of IL-22 in inflammatory response.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CRF2-9; Cytokine receptor class-II member 9; Cytokine receptor family 2 member 9; il-22 receptor; IL-22 receptor subunit alpha-1; IL-22R-alpha-1; IL-22RA1; ILN; Interleukin; interleukin 22 receptor, alpha 1; Interleukin-22 receptor subunit alpha-1; ZcytoR11
Gene Aliases: CRF2-9; IL22R; IL22R1; IL22RA1
UniProt ID: (Human) Q8N6P7
Entrez Gene ID: (Human) 58985
Molecular Function:
transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support