Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Interleukin 24 (IL-24)/melanoma differentiation-associated gene-7 (mda-7) encodes a secreted protein that exhibits significant homology to the interleukin 10 (IL-10) family of cytokines. The human IL-24 is secreted by activated peripheral blood mononuclear cells and binds two heterodimeric receptors, IL-22R1/IL-20R2 and IL-20R1/IL-20R2 resulting in the activation of JAK/STAT signaling pathways. Mda-7/IL-24, selectively induces growth suppression, apoptosis and radiosensitization in malignant gliomas in a p53-independent manner.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cytokine-like protein Mob-5; IL 24; IL-24; IL-4-induced secreted protein; Interleukin-24; MDA-7; melanocyte-associated Mda-7; Melanoma differentiation-associated gene 7 protein; suppression of tumorigenicity 16 (melanoma differentiation); Suppression of tumorigenicity 16 protein; Th2-specific cytokine FISP
Gene Aliases: C49A; FISP; IL10B; IL24; Mda-7; MDA7; MOB5; ST16
UniProt ID: (Human) Q13007, (Rat) Q9JI24, (Mouse) Q925S4
Entrez Gene ID: (Human) 11009, (Rat) 170819, (Mouse) 93672
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support