Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
In direct ELISAs, no cross-reactivity with recombinant mouse (rm) IL-2 R alpha, rmIL-2 R beta, recombinant human (rh) IL-2 R gamma, rhIL-15 R alpha, recombinant rat IL-2 R alpha, or recombinant canine IL-2 R alpha is observed.
Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.
IL2RG is an important signaling component of many interleukin receptors, including those of interleukin -2, -4, -7 and -21, and is thus referred to as the common gamma chain. Mutations in this gene cause X-linked severe combined immunodeficiency (XSCID), as well as X-linked combined immunodeficiency (XCID), a less severe immunodeficiency disorder.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD132; Cytokine receptor common subunit gamma; gammaC; IL-2 receptor subunit gamma; IL2R gamma; Interleukin-2 receptor subunit gamma; p64
Gene Aliases: [g]c; CD132; gamma(c); gc; Il2rg; p64
UniProt ID: (Mouse) P34902
Entrez Gene ID: (Mouse) 16186
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support