Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Sequence of this protein is as follows: MMWTWALWML PSLCKFSLAA LPAKPENISC VYYYRKNLTC TWSPGKETSY TQYTVKRTYA FGEKHDNCTT NSSTSENRAS CSFFLPRITI PDNYTIEVEA ENGDGVIKSH MTYWRLENIA KTEPPKIFRV KPVLGIKRMI QIEWIKPELA PVSSDLKYTL RFRTVNSTSW MEVNFAKNRK DKNQTYNLTG LQPFTEYVIA LRCAVKESKF WSDWSQEKMG MTEEEAPCGL ELWRVLKPAE ADGRRPVRLL WKKARGAPVL EKTLGYNIWY YPESNTNLTE TMNTTNQQLE LHLGGESFWV SMISYNSLGK SPVATLRIPA IQEKSFQCIE VMQACVAEDQ LVVKWQSSAL DVNTWMIEWF PDVDSEPTTL SWESVSQATN WTIQQDKLKP FWCYNISVYP MLHDKVGEPY SIQAYAKEGV PSEGPETKVE NIGVKTVTIT WKEIPKSERK GIICNYTIFY QAEGGKGFSK TVNSSILQYG LESLKRKTSY IVQVMASTSA GGTNGTSINF KTLSFSVFEI ILITSLIGGG LLILIILTVA YGLKKPNKLT HLCWPTVPNP AESSIATWHG DDFKDKLNLK ESDDSVNTED RILKPCSTPS DKLVIDKLVV NFGNVLQEIF TDEARTGQEN NLGGEKNGYV TCPFRPDCPL GKSFEELPVS PEIPPRKSQY LRSRMPEGTR PEAKEQLLFS GQSLVPDHLC EEGAPNPYLK NSVTAREFLV SEKLPEHTKG EV
IL-31 is a T cell cytokine that is preferentially produced by T helper type 2 cells. IL-31 signals through a heterodimeric receptor composed of the IL-31 receptor (IL-31R) and the oncostatin M receptor (OSM). This receptor complex recruits JAK1, JAK2, Stat1, Stat3 and Stat5 signaling pathways, as well as the PI3 kinase/AKT cascade. SHP-2 and Shc adapter molecules are also recruited and contribute to an increased activation of the MAP kinase pathway in response to IL-31. Overexpression of IL-31 in mice results in pruritus and skin dermatitis resembling human atopic dermatitis (AD). Comparisons between skin from patients with AD and healthy skin showed IL-31R expression at higher levels on epidermal keratinocytes in AD samples. Infiltrating cells, more numerous in skin from patients with AD compared with that of healthy individuals, expressed IL-31 mRNA. IL-31 may participate in the cause of itch sensation and promote scratching behavior in NC/Nga mice with atopic dermatitis, and may represent a novel target for antipruritic drug development.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: class I cytokine receptor; CRL 3; CRL3Glmr; Cytokine receptor-like 3; GLM R; GLM-R; GLM-RMGC125346; GP130 like receptor; Gp130-like monocyte receptor; Gp130-like receptor; HGLM R; hGLM-R; HGLMR; IL 31RA; IL-31 receptor subunit alpha; IL-31R subunit alpha; IL-31RAGPLCRL; IL31R subunit alpha; Interleukin-31 receptor subunit alpha; MGC125346; PLCA 2; soluble type I cytokine receptor CRL3; ZcytoR17
Gene Aliases: CRL; CRL3; GLM-R; GLMR; GPL; hGLM-R; IL-31RA; IL31RA; PLCA2; PRO21384; UNQ6368/PRO21073/PRO21384
UniProt ID: (Human) Q8NI17
Entrez Gene ID: (Human) 133396
Molecular Function:
transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support