Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
In direct ELISAs, no cross-reactivity with recombinant human (rh) IL-9 R, recombinant mouse IL-4 R, rhIL-5 R alpha, rhIL-5 R beta, rhIL-13 R alpha 1, or rhIL-13 R alpha 2 is observed.
Reconstitute at 0.5 mg/mL in sterile PBS.
Endoxin level is <0.10 EU per 1 µg of the antibody by the LAL method.
IL4 Receptor alpha (IL4RA) is the alpha chain of the interleukin 4 receptor which binds to both interleukin 4 and interleukin 13 to regulate IgE production, chemokine and mucus production at sites of allergic inflammmation. The IL4 response is involved in promoting Th2 differentiation. The secreted extracellular domain of IL4R alpha, called sIL4R alpha, can inhibit IL4-mediated cell proliferation and IL5 upregulation by T-cells. It has no signaling abilities. Allelic variations in the IL4RA gene have been associated with atopy, a condition that can manifest itself as allergic rhinitis, sinusitus, asthma, or eczema.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD124; il-4 receptor; IL-4 receptor subunit alpha; IL4R nirs variant 1; interleukin-4 receptor alpha chain; Interleukin-4 receptor subunit alpha
Gene Aliases: 582J2.1; CD124; IL-4RA; IL4R; IL4RA
UniProt ID: (Human) P24394
Entrez Gene ID: (Human) 3566
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support