Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
The recommended shelf life for this product is 1 year from date of receipt.
Application Note: For ICC, acetone fixation is recommended. Permeabilization is not recommended. Formaldehyde fixation is not recommended.
Based on 100% sequence identity, this antibody is predicted to react with Rat, Bovine, Guinea pig_10141 and Orangutan
Transduction of extracellular matrix signals through integrins influences intracellular and extracellular functions, and appears to require interaction of integrin cytoplasmic domains with cellular proteins. Integrin-linked kinase (ILK), interacts with the cytoplasmic domain of beta-1 integrin. This gene encodes a serine/threonine protein kinase with 4 ankyrin-like repeats, which associates with the cytoplasmic domain of beta integrins and acts as a proximal receptor kinase regulating integrin-mediated signal transduction. Multiple alternatively spliced transcript variants encoding the same protein have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 59 kDa serine/threonine-protein kinase; Beta-integrin-linked kinase; epididymis secretory protein Li 28; ILK-1; ILK-2; integrin binding protein kinase; Integrin linked ILK; integrin-linked kinase; integrin-linked kinase-2; Integrin-linked protein kinase; p59ILK
Gene Aliases: AA511515; ESTM24; HEL-S-28; ILK; ILK-1; ILK-2; ILK1; ILK2; P59; p59ILK
UniProt ID: (Human) Q13418, (Mouse) O55222
Entrez Gene ID: (Human) 3611, (Mouse) 16202
Molecular Function:
non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support