Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: IGDGQYKRCG VTSVPDIRRC QLTPNDRFIL LACDGLFKVF TPEEAVNFIL SCLEDEKIQT REGKSAADAR YEAACNRLAN KAVQRGSADN VTVMVVRIGH
The protein encoded by this gene is a protein serine/threonine phosphatase of the PP2C family. This protein can interact with integrin-linked kinase (ILK/ILK1), a regulator of integrin mediated signaling, and regulate the kinase activity of ILK. Through the interaction with ILK, this protein may selectively affect the signaling process of ILK-mediated glycogen synthase kinase 3 beta (GSK3beta), and thus participate in Wnt signaling pathway.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EC 3.1.3.16; ILKAP; integrin-linked kinase associated phosphatase; Integrin-linked kinase-associated serine/threonine phosphatase 2C; protein phosphatase 2c, delta isozyme; protein phosphatase, Mg2+/Mn2+ dependent 1O
Gene Aliases: ILKAP; ILKAP2; ILKAP3; PP2C-DELTA; PPM1O
UniProt ID: (Human) Q9H0C8
Entrez Gene ID: (Human) 80895
Molecular Function:
protein phosphatase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support