Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Antibody detects endogenous levels of total IMMP2L.
IMP2L complex generates mature, active proteins in the mitochondrial intermembrane space by proteolytically removing the mitochondrial targeting presequence of nuclear-encoded proteins. IMP1 and IMP2 are the catalytic subunits of the IMP complex.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: IMP2 inner mitochondrial membrane peptidase-like; IMP2 inner mitochondrial membrane peptidase-like (S. cerevisiae); IMP2-like protein; inner mitochondrial membrane peptidase 2 like; Mitochondrial inner membrane protease subunit 2
Gene Aliases: AI853880; IMMP2L; IMMP2L-IT1; IMP2; IMP2-LIKE
UniProt ID: (Human) Q96T52, (Mouse) Q8BPT6
Entrez Gene ID: (Human) 83943, (Mouse) 93757, (Rat) 100359529
Molecular Function:
protease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support