Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This gene encodes the human homolog of the yeast Imp3 protein. The protein localizes to the nucleoli and interacts with the U3 snoRNP complex. The protein contains an S4 domain.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BRMS2; DKFZp586L0118; FLJ10968; IMP3, U3 small nucleolar ribonucleoprotein, homolog; insulin-l; OTTHUMP00000183524; U3 small nucleolar ribonucleoprotein protein IMP3; U3 snoRNP protein 3 homolog; U3 snoRNP protein IMP3; VICKZ3
Gene Aliases: 1190002L16Rik; AI256594; BRMS2; C15orf12; IMP3; MRPS4
UniProt ID: (Human) Q9NV31, (Mouse) Q921Y2
Entrez Gene ID: (Human) 55272, (Mouse) 102462
Molecular Function:
ribosomal protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support