Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MLGQQQQQL YSSAALLTGE RSRLLTCYVQ DYLECVESLP HDMQRNVSVL RELDNKYQET LKEIDDVYEK YKKEDDLNQK KRLQQLLQRA LINSQELGDE KIQIVTQMLE LVENRARQME LHSQCFQDPA ESERASDKAK MDSSQPERSS RRPRRQRTSE SRDLCHMANG IEDCDDQPPK EKKSKSAKKK KRSKAKQERE ASPVEFAIDP NEPTYCLCNQ VSYGEMIGCD NEQCPIEWFH FSCVSLTYKP KGKWYCPKCR GDNEKTMDKS TEKTKKDRRS R (1-280 aa encoded by BC030128)
This gene is a member of the inhibitor of growth (ING) family. Members of the ING family associate with and modulate the activity of histone acetyltransferase (HAT) and histone deacetylase (HDAC) complexes and function in DNA repair and apoptosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ING1Lp; Inhibitor of growth 1-like protein; inhibitor of growth family, member 2; Inhibitor of growth protein 2; p32; p33ING2
Gene Aliases: 2810011M06Rik; ING1L; ING2; Ing2b; P33ING2
UniProt ID: (Human) Q9H160, (Mouse) Q9ESK4
Entrez Gene ID: (Human) 3622, (Mouse) 69260, (Rat) 290744
Molecular Function:
chromatin/chromatin-binding, or -regulatory protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support