Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Positive Samples: Mouse liver, Mouse lung
Immunogen sequence: MKGGFTGGDE YQKHFLPRDY LATYYSFDGS PSPEAEMLKF NLECLHKTFG PGGLQGDTLI DIGSGPTIYQ VLAACDSFQD ITLSDFTDRN REELEKWLKK EPGAYDWTPA VKFACELEGN SGRWEEKEEK LRAAVKRVLK CDVHLGNPLA PAVLPLADCV LTLLAMECAC CSLDAYRAAL CNLASLLKPG GHLVTTVTLR LPSYMVGKRE FSCVALEKEE VEQAVLDAGF DIEQLLHSPQ SYSVTNAANN GVCFIVARKK PGP
N-methylation of endogenous and xenobiotic compounds is a major method by which they are degraded. This gene encodes an enzyme that N-methylates indoles such as tryptamine.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Amine N-methyltransferase; Aromatic alkylamine N-methyltransferase; arylamine N-methyltransferase; Indolamine N-methyltransferase; Indolethylamine N-methyltransferase; nicotine N-methyltransferase; TEMT; Thioether S-methyltransferase
Gene Aliases: INMT; TEMT
UniProt ID: (Human) O95050, (Mouse) P40936
Entrez Gene ID: (Human) 11185, (Mouse) 21743
Molecular Function:
methyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support