Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
DNA helicase and probable main scaffold component of the chromatin remodeling INO80 complex which is involved in transcriptional regulation, DNA replication and probably DNA repair; according to PubMed:20687897 the contribution to DNA double-strand break repair appears to be largely indirect through transcriptional regulation. Recruited by YY1 to YY1-activated genes, where it acts as an essential coactivator. Binds DNA. In vitro, has double-stranded DNA-dependent ATPase activity. Involved in UV-damage excision repair, DNA replication and chromosome segregation during normal cell division cycle.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Chromatin-remodeling ATPase INO80; DNA helicase INO80; DNA helicase-related INO80 complex homolog 1; DNA helicase-related protein INO80; hINO80; homolog of yeast INO80; INO80 complex homolog 1; INO80 complex subunit A; INO80 homolog; putative DNA helicase INO80 complex homolog 1; yeast INO80-like protein
Gene Aliases: 2310079N15Rik; 4632409L19Rik; hINO80; INO80; INO80A; INOC1; KIAA1259; RGD1310969
UniProt ID: (Human) Q9ULG1, (Mouse) Q6ZPV2
Entrez Gene ID: (Human) 54617, (Mouse) 68142, (Rat) 296084
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support