Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Knockout to ensure that the antibody binds to the antigen stated.
Peptide sequence: VVKLIFRESD NDRKVMLQLE KKLFDYFNQE VFRDNNGTAL LEFDKELSVF
Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 93%; Pig: 100%; Rat: 93%
The protein encoded by this gene is a membrane-associated type I inositol 1,4,5-trisphosphate (InsP3) 5-phosphatase. InsP3 5-phosphatases hydrolyze Ins(1,4,5)P3, which mobilizes intracellular calcium and acts as a second messenger mediating cell responses to various stimulation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 43 kDa inositol polyphosphate 5-phophatase; 43 kDa inositol polyphosphate 5-phosphatase; 5PTase; CTCL tumor antigen HD-CL-02; DKFZp434A1721; Inositol polyphosphate-5-phosphatase A; inositol polyphosphate-5-phosphatase, 40kD; inositol polyphosphate-5-phosphatase, 40kDa; inositol trisphosphate-5-phosphatase, 40kD; InsP3 5-phosphatase; MGC116947; MGC116949; Type I inositol 1,4,5-trisphosphate 5-phosphatase; type I inositol-1,4,5-trisphosphate 5-phophatase; type I inositol-1,4,5-trisphosphate 5-phosphatase
Gene Aliases: 5PTASE; INPP5A
UniProt ID: (Human) Q14642
Entrez Gene ID: (Human) 3632
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support