Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: SPLSFIKVS HVRMQGILLL VFAKYQHLPY IQILSTKSTP TGLFGYWGNK GGVNICLKLY GYYVSIINCH LPPHISNNYQ RLEHFDRILE MQNCEGRDIP NILDHDLIIW FGDMNFRIED FGLHFVRESI KNRCYGGLWE KDQLSIAKKH DPLLREFQEG RLLFPPTYKF DRNSNDYDTS EKKRKPAWTD RILWRLKRQP CAGPDTPIPP ASHFSLSLRG YSSHMTYGIS DHKPVSGTFD LELKPLVSAP LIVLMPEDLW TVENDMMVSY SSTSDFPSSP WDWIGLYKVG LRDVNDYVSY AWVGDSKVSC SDNLNQVYID ISNIPTTEDE FLLCYYSNSL RSVVGISRPF QIPPGSLRED PLGEAQPQI (81-448 aa encoded by BC004362)
This gene encodes a protein with 5-phosphatase activity toward polyphosphate inositol. The protein localizes to the cytosol in regions lacking actin stress fibers. It is thought that this protein may negatively regulate the actin cytoskeleton. Alternatively spliced transcript variants encoding different isoforms have been identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C62; Inositol polyphosphate 5-phosphatase K; Phosphatidylinositol-3,4,5-trisphosphate 5-phosphatase; Phosphatidylinositol-4,5-bisphosphate 5-phosphatase; PI-5-phosphatase related; putative phosphoinositide 5-phosphatase type II; putative phosphoinositide 5-phosphatase type II; C62; putative PI-5-phosphatase; skeletal muscle and kidney enriched inositol phosphatase; skeletal muscle and kidney enriched inositol polyphosphate phosphatase; Skeletal muscle and kidney-enriched inositol phosphatase
Gene Aliases: C62; INPP5K; PPS; SKIP
UniProt ID: (Human) Q9BT40, (Mouse) Q8C5L6
Entrez Gene ID: (Human) 51763, (Rat) 287533, (Mouse) 19062
Molecular Function:
phosphatase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support