Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Tissue lysate).
Immunogen sequence: MSSRKLSGPK GRRLSIHVVT WNVASAAPPL DLSDLLQLNN RNLNPDIYVI GLQELNSGII SLLSDAAFND SWSSFLMDVL SPLSFIKVSH VRMQGILLLV FAKYQHLPYI QILSTKSTPT GLFGYWGNKG GVNICLKLYG YYVSIINCHL PPHISNNYQR LEHFDRILEM QNCEGRDIPN ILDHDLIIWF GDMNFRIEDF GLRFVRESIK NRCYGGLWEK DQLSIAKKHD PLLREFQEGR LLFPPTYKFD RNSNDYDTSE KKRKPAWTDR ILWRLKRQPC AGPDTPIPPA SHFSLSLRGY SSHMTYGISD HKPVSGTFDL ELKPLVSAPL IVLMPEDLWT VENDMMVSYS STSDFPSSPW DWIGLYKVGL RDVNDYVSYA WVGDSKVSCS DNLNQVYIDI SNIPTTEDEF LLCYYSNSLR SVVGISRPFQ IPPGSLREDP LGEAQPQI
This gene encodes a protein with 5-phosphatase activity toward polyphosphate inositol. The protein localizes to the cytosol in regions lacking actin stress fibers. It is thought that this protein may negatively regulate the actin cytoskeleton. Alternatively spliced transcript variants encoding different isoforms have been identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Inositol polyphosphate 5-phosphatase K; Phosphatidylinositol-3,4,5-trisphosphate 5-phosphatase; Phosphatidylinositol-4,5-bisphosphate 5-phosphatase; Skeletal muscle and kidney-enriched inositol phosphatase
Gene Aliases: INPP5K; PPS; SKIP
UniProt ID: (Human) Q9BT40
Entrez Gene ID: (Human) 51763
Molecular Function:
phosphatase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support