Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: LYLNDYCHRG LRLPTSNNDP RFDGEDGDPE AEMESDCCPC QHPPPGQVLP PLEAQEASFQ KKFENFLHNA ITIPISPWKV TSINKSPQRD SGRHRRAAGP LRLGGNSSDF
INSRR, insulin receptor-related receptor, is a member of the insulin receptor (IR) family. INSRR does not bind insulin-like molecules, but is similar to INSR in tyrosine kinase activity. Receptor with tyrosine-protein kinase activity. Functions as a pH sensing receptor which is activated by increased extracellular pH. Activates an intracellular signaling pathway that involves IRS1 and AKT1/PKB.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Insulin receptor-related protein; insulin receptor-related receptor; IR-related receptor; IRR
Gene Aliases: INSRR; IRR; Sirr
UniProt ID: (Human) Q5VZS3, (Rat) Q64716
Entrez Gene ID: (Human) 3645, (Rat) 60663
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support