Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MASVASCDS RPSSDELPGD PSSQEEDEDY DFEDRVSDSG SYSSASSDYD DLEPEWLDSV QKNGELFYLE LSEDEEESLL PETPTVNHVR FSENEIIIED DYKERKKYEP KLKQFTKILR RKRLLPKRCN KKNSNDNGPV SILKHQSNQK TGVIVQQRYK DVNVYVNPKK LTVIKAKEQL KLLEVLVGII HQTKWSWRRT GKQGDGERLV VHGLLPGGSA MKSGQVLIGD VLVAVNDVDV TTENIERVLS CIPGPMQVKL TFENAYDVKR ETSHPRQKKT QSNTSDLVKL LWGEEVEGIQ QSGLNTPHII MYLTLQLDSE TSKEEQEILY HYPMSEASQK LKSVRGIFLT LCDML (1-354 aa encoded by BC130611)
Plays a key role in ciliogenesis and embryonic development. Regulator of cilia formation by controlling the organization of the apical actin cytoskeleton and the positioning of the basal bodies at the apical cell surface, which in turn is essential for the normal orientation of elongating ciliary microtubules. Plays a key role in definition of cell polarity via its role in ciliogenesis but not via conversion extension. Has an indirect effect on hedgehog signaling. Proposed to function as core component of the CPLANE (ciliogenesis and planar polarity effectors) complex involved in the recruitment of peripheral IFT-A proteins to basal bodies (PubMed:27158779). [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: homolog of inturned; Inturned planar cell polarity effector homolog; PDZ domain containing 6; PDZ domain-containing protein 6; Protein inturned
Gene Aliases: 9230116I04Rik; 9430087H23Rik; INT; INTU; KIAA1284; mKIAA1284; PDZD6; PDZK6
UniProt ID: (Human) Q9ULD6, (Mouse) Q059U7
Entrez Gene ID: (Human) 27152, (Mouse) 380614
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support