Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: IQ domain-containing protein C; IQ motif containing C; RP4-622L5.6
Gene Aliases: BC026978; IQCC
UniProt ID: (Human) Q4KMZ1, (Mouse) A2ADZ8
Entrez Gene ID: (Human) 55721, (Rat) 682431, (Mouse) 230767
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support