Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: GQRAVKTHG ARTKYLVYER LEKLQAVVAG VPGHLEAASC IPPSPQENSY VSSTGRAHSG AAPWQPLAAP SGASAQAAEQ LQRGPNQPVE SDESLGGLSA ALRSWHLTPS CPLDPAPLRE AGCPQGDTAG ESSWGSGPGS RPTAVEGLAL GSSASSSSEP PQIIINPARQ KMVQKLALYE DGALDSLQLL SSSSLPGLGL EQDRQGPEES DEFQS (420-633 aa encoded by BC014963)
IL-1 receptor associated kinases (IRAKs) are serine/threonine kinases involved in signaling via Toll/IL-1 pathways. It is rapidly recruited by MYD88 to the receptor-signaling complex upon TLR activation. IRAK's association with MYD88 leads to IRAK1 phosphorylation by IRAK4 and subsequent autophosphorylation and kinase activation. Alternatively, IRAK1 can phosphorylate TIRAP to promote its ubiquitination and subsequent degradation. IRAK1 also phosphorylates the interferon regulatory factor 7 (IRF7) to induce its activation and translocation to the nucleus. This results in the transcriptional activiation of type I IFN genes, which drive the cell in an antiviral state.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Interleukin-1 receptor-associated kinase 1; IRAK-1; Pelle homolog
Gene Aliases: IRAK; IRAK1; pelle
UniProt ID: (Human) P51617
Entrez Gene ID: (Human) 3654
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support