Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: KPEKPLAASV RKAEDEQEEG QPVRMATFPG PGSSPARAHQ PAFLQPPEED APHSLRSDLP TSSDSKDFST SIPKQEKLLS LAGDSLFWSE ADVVQATDDF
IRAK2 (interleukin-1 receptor-associated kinase-like 2) binds to the IL-1 type I receptor following IL-1 engagment and triggers intracellular signaling cascades leading to transcriptional up-regulation and mRNA stabilization. The pro-inflammatory cytokine IL-1 induces cellular response through two subunits of its receptor, IL-1 receptor I (IL-1RI) and IL-1 receptor accessory protein (IL-1RAcP). IL-1 receptor-associated kinase (IRAK) mediates activation of NF-kappa-B, which is a pivotal transcription factor mediating inflammatory and immune response. A novel member in the IRAK/Pelle family was recently identified and designated IRAK2. Both IRAK and IRAK2 recruit to the subunits of the IL-1R complex after IL-1 binding and lead to NF-kappa-B activation. IRAKs also associate with Toll-like receptor (TLR) and the dominant negative mutants of IRAKs inhibit LPS-induced NF-kappa-B activation. Members in IRAK/Pelle family play a central role in IL-1R and TLR mediated inflammatory responses to cytokine IL-1 and LPS. IRAK2 is expressed in a variety of human tissues.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: interleukin-1 receptor associated kinase-2; interleukin-1 receptor-associated kinase 2; interleukin-1 receptor-associated kinase 2c; Interleukin-1 receptor-associated kinase-like 2; IRAK-2; MGC150550; mu-IRAK-2
Gene Aliases: 6330415L08Rik; AI649099; IRAK-2; IRAK2
UniProt ID: (Human) O43187, (Mouse) Q8CFA1
Entrez Gene ID: (Human) 3656, (Mouse) 108960
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support