Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: RIDRMTQKTP FECSQSEVMF LSLDKKPESK RNEEACNMPS SSCEESWFPK YIVPSQDLRP YKVNIDPSSE APGHSCRSRP VESSCSSKFS WDEYEQYKKE
IRAK3 serine/threonine kinase or interleukin-1 receptor-associated kinase 3 is a member of the interleukin-1 receptor-associated kinase protein family which are essential components of the Toll/IL-R immune signal transduction pathways. IRAK3 is primarily expressed in monocytes and macrophages and functions as a negative regulator of Toll-like receptor signaling. IRAK3 mediates critical aspects of innate immunity that result in an immunocompromised state during sepsis. Mutations in IRAK3 are associated with a susceptibility to asthma.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ13601; IL-1 receptor-associated kinase M; Inactive IL-1 receptor-associated kinase 3; Interleukin-1 receptor-associated kinase 3; IRAK-3; IRAK-M; IRAK3
Gene Aliases: ASRT5; IRAK3; IRAKM
UniProt ID: (Human) Q9Y616
Entrez Gene ID: (Human) 11213
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support