Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: MNKPITPSTY VRCLNVGLIR KLSDFIDPQE GWKKLAVAIK KPSGDDRYNQ FHIRRFEALL QTGKSPTSEL LFDWGTTNCT VGDLVDLLIQ NEFFAPASLL LPDAVPKTAN TLPSKEAITV QQKQMPFCDK DRTLMTPVQN LEQSYMPPDS SSPENKSLEV SDTRFHSFSF YELKNVTNNF DERPISVGGN KMGEGGFGVV
IL-1 receptor associated kinases (IRAKs) are serine/threonine kinases involved in signaling via Toll/IL-1 pathways. Overexpression of IRAK-4 activates NF-kappa-B and MAPK pathways. IRAK-4 interacts with and phosphorylates IRAK-1. IRAK-4-deficient animals are completely resistant to the challenge with LPS. Animals and humans lacking IRAK-4 are impaired in their responses to viral and bacterial challenges. Members in IRAK/Pelle family play a central role in IL-1R/TLR mediated inflammatory responses and in innate immunity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: interleukin-1; Interleukin-1 receptor-associated kinase 4; IRAK-4; Renal carcinoma antigen NY-REN-64
Gene Aliases: IPD1; IRAK-4; IRAK4; NY-REN-64; REN64
UniProt ID: (Human) Q9NWZ3
Entrez Gene ID: (Human) 51135
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support