Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: PQKPEDKADP EGPEAGGAEQ KAASGCERLQ GPPTPAGKET EGSLSDSDFK EPPSEGRLDA L
Establishes the cardiac repolarization gradient by its repressive actions on the KCND2 potassium-channel gene. Required for retinal cone bipolar cell differentiation. May regulate contrast adaptation in the retina and control specific aspects of visual function in circuits of the mammalian retina. Could be involved in the regulation of both the cell cycle and apoptosis in prostate cancer cells. Involved in craniofacial and gonadal development. Modulates the migration of progenitor cell populations in branchial arches and gonads by repressing CXCL12.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Homeodomain protein IRX-2A; Homeodomain protein IRXB2; Iroquois homeobox protein 5; Iroquois-class homeodomain protein IRX-5
Gene Aliases: HMMS; IRX-2a; IRX2A; IRX5; IRXB2
UniProt ID: (Human) P78411
Entrez Gene ID: (Human) 10265
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support