Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAGSREVVAM DCEMVGLGPH RESGLARCSL VNVHGAVLYD KFIRPEGEIT DYRTRVSGVT PQHMVGATPF AVARLEILQL LKGKLVVGHD LKHDFQALKE DMSGYTIYDT STDRLLWREA KLDHCRRVSL RVLSERLLHK SIQNSLLGHS SVEDARATME LYQISQRIRA RRGLPRLAVS D
Interferon-induced antiviral exoribonuclease that acts on single-stranded RNA and also has minor activity towards single-stranded DNA. Exhibits antiviral activity against RNA viruses including hepatitis C virus (HCV), hepatitis A virus (HAV) and yellow fever virus (YFV) in an exonuclease-dependent manner. May also play additional roles in the maturation of snRNAs and rRNAs, and in ribosome biogenesis. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Estrogen-regulated transcript 45 protein; interferon stimulated exonuclease gene 20kDa; Interferon-stimulated gene 20 kDa protein; Promyelocytic leukemia nuclear body-associated protein ISG20
Gene Aliases: CD25; HEM45; ISG20
UniProt ID: (Human) Q96AZ6
Entrez Gene ID: (Human) 3669
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support