Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Sequence of this protein is as follows: MGDPPKKKRL ISLCVGCGNQ IHDQYILRVS PDLEWHAACL KCAECNQYLD ESCTCFVRDG KTYCKRDYIR LYGIKCAKCS IGFSKNDFVM RARSKVYHIE CFRCVACSRQ LIPGDEFALR EDGLFCRADH DVVERASLGA GDPLSPLHPA RPLQMAAEPI SARQPALRPH VHKQPEKTTR VRTVLNEKQL HTLRTCYAAN PRPDALMKEQ LVEMTGLSPR VIRVWFQNKR CKDKKRSIMM KQLQQQQPND KTNIQGMTGT PMVAASPERH DGGLQANPVE VQSYQPPWKV LSDFALQSDI DQPAFQQLVN FSEGGPGSNS TGSEVASMSS QLPDTPNSMV ASPIEA
This gene encodes a member of the LIM/homeodomain family of transcription factors. The encoded protein binds to the enhancer region of the insulin gene, among others, and may play an important role in regulating insulin gene expression. The encoded protein is central to the development of pancreatic cell lineages and may also be required for motor neuron generation. Mutations in this gene have been associated with maturity-onset diabetes of the young.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Insulin gene enhancer protein ISL-1; insulin related protein; isl-1 homeobox; ISL1 transcription factor LIM/homeodomain (islet-1); ISL1 transcription factor, LIM/homeodomain; ISL1 transcription factor, LIM/homeodomain 1; Islet 1; Islet-1; Lim-homeodomain protein Islet1
Gene Aliases: Isl-1; isl-1=homeobox; ISL1; ISLET1
UniProt ID: (Human) P61371, (Mouse) P61372, (Rat) P61374
Entrez Gene ID: (Human) 3670, (Mouse) 16392, (Rat) 64444
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support