Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MVDIIFHYP FLGAMGDHSK KKPGTAMCVG CGSQIHDQFI LRVSPDLEWH AACLKCAECS QYLDETCTCF VRDGKTYCKR DYVRLFGIKC AKCQVGFSSS DLVMRARDSV YHIECFRCSV CSRQLLPGDE FSLREHELLC RADHGLLLER AAAGSPRSPG PLPGARGLHL PDAGSGRQPA LRPHVHKQTE KTTRVRTVLN EKQLHTLRTC YAANPRPDAL MKEQLVEMTG LSPRVIRVWF QNKRCKDKKK SILMKQLQQQ QHSDKTSLQG LTGTPLVAGS PIRHENAVQG SAVEVQTYQP PWKALSEFAL QSDLDQPAFQ QLVSFSESGS LGNSSGSDVT SLSSQLPDTP NSMVPSPVET (1-359 aa encoded by BC012136)
ISL2 is a transcriptional factor that defines subclasses of motoneurons that segregate into columns in the spinal cord and select distinct axon pathways.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Insulin gene enhancer protein ISL-2; insulin related protein 2 (islet 2); ISL2 transcription factor, LIM/homeodomain, (islet-2); islet 2; Islet-2
Gene Aliases: 3110001N10Rik; Isl-2; ISL2
UniProt ID: (Human) Q96A47, (Mouse) Q9CXV0, (Rat) P50480
Entrez Gene ID: (Human) 64843, (Mouse) 104360, (Rat) 57233
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support