Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: MLGSGFKAER LRVNLRLVIN RLKLLEKKKT ELAQKARKEI ADYLAAGKDE RARIRVEHII REDYLVEAME ILELYCDLLL ARFGLIQSMK ELDSGLAESV STLIWAAPRL QSEVAELKIV ADQLCAKYSK EYGKLCRTNQ IGTVNDRLMH KLSVEAPPKI LVERYLIEIA KNYNVPYEPD SVVMAEAPPG VETDLIDVGF TDDVKKGGPG RGGSGGFTAP VGGPDGTVPM PMPMPMPMPS ANTPFSYPLP KGPVDDINAD KNISSAQIVG PGPKPEASAK LPSRPADNYD NFVLPELPSV PDTLPTASAG ASTSASEDID FDDLSRRFEE LKKKT; Positive Samples: A-549, HeLa, Mouse heart, Rat brain; Cellular Location: Cytoplasm, Cytoplasmic vesicle, Midbody, centrosome, cytoskeleton, microtubule organizing center
Proposed to be involved in specific functions of the ESCRT machinery. Is required for efficient abscission during cytokinesis, but not for HIV-1 budding. The involvement in the MVB pathway is not established. Involved in recruiting VPS4A and/or VPS4B to the midbody of dividing cells.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Charged multivesicular body protein 8; CHMP8; hIST1; increased sodium tolerance 1 homolog; IST1 homolog; IST1, endosomal sorting complex required for transport-III component; MAPK activating protein PM28; MGC117220; overexpressed in lung cancer 1; pm28; Putative MAPK-activating protein PM28
Gene Aliases: 2400003C14Rik; AW536298; IST1; KIAA0174; mKIAA0174; OLC1; RGD1307799
UniProt ID: (Human) P53990, (Rat) Q568Z6, (Mouse) Q9CX00
Entrez Gene ID: (Human) 9798, (Rat) 307833, (Mouse) 71955
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support