Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: RDIRDFRSS AGLDKVIVLW TANTERFCEV IPGLNDTAEN LLRTIELGLE VSPSTLFAVA SILEGCAFLN GSPQNTLVPG ALELAWQHRV FVGGDDFKSG QTKVKSVLVD FLIGSGLKTM SIVSYNHLGN NDGENLSAPL QFRSKEVSKS NVVDDMVQSN QVLYTPGEEP DHCVVIKYVP YVGDGKRALD EYTSELMLGG TNTLVLHNTC EDSLLAAPIM LDLALLTELC QRVSFCTDMD PEPQTFHPVL SLLSFLFKAP LVPRGSPVVN ALFRQRSCIE NILRACVGLP PQNHMLLEHK MERPGPSLKR VGPVAATYPM LNKKGPVPAA TNGCTGDANG HLQEEPPMPT T (209-558 aa encoded by BC066902)
Myoinositol, the most common naturally occurring form of inositol, is a component of plasma membrane phospholipids and functions as a cell signaling molecule. ISYNA1 (EC 5.5.1.4), or IPS, is a rate-limiting enzyme that catalyzes the de novo synthesis of myoinositol 1-phosphate from glucose 6-phosphate. Myoinositol, the most common naturally occurring form of inositol, is a component of plasma membrane phospholipids and functions as a cell signaling molecule. ISYNA1 (EC 5.5.1.4), or IPS, is a rate-limiting enzyme that catalyzes the de novo synthesis of myoinositol 1-phosphate from glucose 6-phosphate (Seelan et al., 2004 ).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hINO1; Inositol-3-phosphate synthase 1; IPS 1; MI-1-P synthase; MIP synthase; Myo-inositol 1-phosphate synthase; Myo-inositol 1-phosphate synthase A1; myo-inositol-1-phosphate synthase; testis secretory sperm-binding protein Li 200a
Gene Aliases: 1300017C10Rik; AU018670; INO1; INOS; IPS; IPS 1; IPS-1; ISYNA1
UniProt ID: (Human) Q9NPH2, (Rat) Q6AYK3, (Mouse) Q9JHU9
Entrez Gene ID: (Human) 51477, (Rat) 290651, (Mouse) 71780
Molecular Function:
isomerase
metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support