Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Peptide sequence: LQEQLWPHME ALRPRPSVYI PEFIAANQSA RADNLIPGSR AQQLEQIRRD
Sequence homology: Cow: 86%; Guinea Pig: 93%; Horse: 86%; Human: 100%; Mouse: 86%; Rat: 86%; Yeast: 92%
Myoinositol, the most common naturally occurring form of inositol, is a component of plasma membrane phospholipids and functions as a cell signaling molecule. ISYNA1 (EC 5.5.1.4), or IPS, is a rate-limiting enzyme that catalyzes the de novo synthesis of myoinositol 1-phosphate from glucose 6-phosphate. Myoinositol, the most common naturally occurring form of inositol, is a component of plasma membrane phospholipids and functions as a cell signaling molecule. ISYNA1 (EC 5.5.1.4), or IPS, is a rate-limiting enzyme that catalyzes the de novo synthesis of myoinositol 1-phosphate from glucose 6-phosphate (Seelan et al., 2004 ).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hINO1; Inositol-3-phosphate synthase 1; IPS 1; MI-1-P synthase; MIP synthase; Myo-inositol 1-phosphate synthase; Myo-inositol 1-phosphate synthase A1; testis secretory sperm-binding protein Li 200a
Gene Aliases: INO1; INOS; IPS; IPS 1; IPS-1; ISYNA1
UniProt ID: (Human) Q9NPH2
Entrez Gene ID: (Human) 51477
Molecular Function: isomerase metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support