Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: QFYSSASEYH ISIAANETVP EVINSTEDIG NEINIFYLIR KSGSFPMPEL KLSISFPNMT SNGYPVLYPT GLSSSENANC RPHIFEDPFS INSGKKMTTS TDHLKRGTI
Integrin alpha 1 (ITGA1) chain associates with the beta 1 (ITGB1) chain to form a heterodimer that functions as a dual laminin/collagen receptor in neural cells and hematopoietic cells. ITGA1 has a 206-amino acid I domain in its N-terminal half, followed by 3 divalent cation-binding sites and a C-terminal transmembrane domain with a short cytoplasmic tail. It also has 28 potential N-glycosylation sites. Human ITGA1 was expressed in a mouse fibroblast cell line as a 180-kD protein. ITGA1 is involved in the early remodeling of osteoarthritic cartilage and plays an essential role in the regulation of mesenchymal stem cell proliferation and cartilage production. It also plays an essential role in the regulation of MSC proliferation and cartilage production.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD49 antigen-like family member A; CD49a; integrin alpha 1; Integrin alpha-1; integrin, alpha 1; Laminin and collagen receptor; very late activation protein 1; VLA-1
Gene Aliases: CD49a; ITGA1; VLA1
UniProt ID: (Human) P56199
Entrez Gene ID: (Human) 3672
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support