Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Relative expression to ensure that the antibody binds to the antigen stated.
FIGURE: 1 / 1
Immunogen sequence: HKEEEVGPLV EHIYELHNIG PSTISDTILE VGWPFSARDE FLLYIFHIQT LGPLQCQPNP NINPQDIKPA ASPEDTPELS AFLRNSTIPH LVRKRDVHVV EFHRQSPAKI LNCTNIECLQ ISCA
Integrin alpha-8/beta-1 functions in the genesis of kidney and probably of other organs by regulating the recruitment of mesenchymal cells into epithelial structures. It recognizes the sequence R-G-D in a wide array of ligands including TNC, FN1, SPP1 TGF.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: integrin alpha 8; Integrin alpha-8; integrin, alpha 8
Gene Aliases: ITGA8
UniProt ID: (Human) P53708
Entrez Gene ID: (Human) 8516
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support