Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MVALPMVLVL LLVLSRGESE LDAKIPSTGD ATEWRNPHLS MLGSCQPAPS CQKCILSHPS CAWCKQLNFT ASGEAEARRC ARREELLARG CPLEELEEPR GQQEVLQDQP LSQGARGEGA TQLAPQRVRV TLRPGEPQQL QVRFLRAEGY PVDLYYLMDL SYSMKDDLER VRQLGHALLV RLQEVTHSVR IGFGSFVDKT VLPFVSTVPS KLRHPCPTRL ERCQSPFSFH HVLSLTGDAQ AFEREVGRQS VSGNLDSPEG GFDAILQAAL CQEQIGWRNV SRLLVFTSDD TFHTAGDGKL GGIFMPSDGH CHLDSNGLYS RSTEFDYPSV GQVAQALSAA NIQPIFAVTS AALPVYQELS KLIPKSAVGE LSEDSSNVVQ LIMDAYNSLS STVTLEHSSL PPGVHISYES QCEGPEKREG KAEDRGQCNH VRINQTVTFW VSLQATHCLP EPHLLRLRAL GFSEELIVEL HTLCDCNCSD TQPQAPHCSD GQGHLQCGVC SCAPGRLGRL CECSVAELSS PDLESGCRAP NGTGPLCSGK GHCQCGRCSC SGQSSGHLCE CDDASCERHE GILCGGFGRC QCGVCHCHAN RTGRACECSG DMDSCISPEG GLCSGHGRCK CNRCQCLDGY YGALCDQCPG CKTPCERHRD CAECGAFRTG PLATNCSTAC AHTNVTLALA PILDDGWCKE RTLDNQLFFF LVEDDARGTV VLRVRPQEKG ADHTQAIVLG CVGGIVAVGL GLVLAYRLSV EIYDRREYSR FEKEQQQLNW KQDSNPLYKS AITTTINPRF QEADSPTL
Integrin beta 7 associates with integrin alpha 4 (CD49d) to form the alpha 4 beta 7 integrin LPAM-1, expressed on intraepithelial lymphocytes. It also associates with alpha E (CD103) to form the alpha E beta 7 integrin HML-1, expressed on T cells adjacent to mucosal epithelium and intraepithelial lymphocytes. Ligands for LPAM-1 include VCAM-1 (CD106), MAdCAM-1 and fibronectin, while the ligand of HML-1 is E-cadherin (CD324). Integrin beta 7 plays an important role in the adhesion of leukocytes to endothelial cells promoting the transmigration of leukocytes to extravascular spaces during the inflammatory response.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Gut homing receptor beta subunit; integrin beta 7 subunit; Integrin beta-7; integrin, beta 7
Gene Aliases: ITGB7
UniProt ID: (Human) P26010
Entrez Gene ID: (Human) 3695
Molecular Function:
integrin
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support