Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: LLSDSESRS RGNVQLEAGE DVGQKNHWQK IRTMVNLPVI SPFKKRYAWV QLAGHTGSFK AAGTSGLILK RCSEPERYCL ARLMADALRG CVPAFHGVVE RDGESYLQLQ DLLDGFDGPC VLDCKMGVRT YLEEELTKAR ERPKLRKDMY KKMLAVDPEA PTEEEHAQRA VTKPRYMQWR EGISSSTTLG FRIEGIKKAD GSCSTDFKTT RSREQVLRVF EEFVQGDEEV LRRYLNRLQQ IRDTLEVSEF FRRHEVIGSS LLFVHDHCHR AGVWLIDFGK TTPLPDGQIL DHRRPWEEGN REDGYLLGLD NLIGILASLA ER (141-461 aa encoded by BC026331)
Regulates inositol phosphate metabolism by phosphorylation of second messenger inositol 1,4,5-trisphosphate to Ins(1,3,4,5)P4. The activity of the inositol 1,4,5-trisphosphate 3-kinase is responsible for regulating the levels of a large number of inositol polyphosphates that are important in cellular signaling. Both calcium/calmodulin and protein phosphorylation mechanisms control its activity. It is also a substrate for the cyclic AMP-dependent protein kinase, calcium/calmodulin- dependent protein kinase II, and protein kinase C in vitro.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EC 2.7.1.127; inositol 1,4,5-triphosphate 3-kinase; Inositol 1,4,5-trisphosphate 3-kinase A; inositol 145-triphosphate 3-kinase; Inositol-trisphosphate 3-kinase A; insP 3-kinase A; IP3 3 kinase A; IP3 3-kinase A; IP3-kinase A; IP3K A
Gene Aliases: IP3-3KA; IP3KA; ITPKA
UniProt ID: (Human) P23677, (Mouse) Q8R071, (Rat) P17105
Entrez Gene ID: (Human) 3706, (Mouse) 228550, (Rat) 81677
Molecular Function:
kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support