Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MRRCPCRGS LNEAEAGALP AAARMGLEAP RGGRRRQPGQ QRPGPGAGAP AGRPEGGGPW ARTEGSSLHS EPERAGLGPA PGTESPQAEF WTDGQTEPAA AGLGVETERP KQKTEPDRSS LRTHLEWSWS ELETTCLWTE TGTDGLWTDP HRSDLQFQPE EASPWTQPGV HGPWTELETH GSQTQPERVK SWADNLWTHQ NSSSLQTHPE GACPSKEPSA DGSWKELYTD GSRTQQDIEG PWTEPYTDGS QKKQDTEAAR KQPGTGGFQI QQDTDGSWTQ PSTDGSQTAP GTDCLLGEPE DGPLEEPEPG ELLTHLYSHL KCSPLCPVPR LIITPETPEP EAQPVGPPSR VEGGSGGFS (1-358 aa encoded by BC060788 )
Inositol 1, 4, 5-trisphosphate 3-kinase (ITPK) catalyzes the phosphorylation of Ins(1, 4, 5)P3 to Ins(1, 3, 4, 5)P4, both of which are modulators of calcium homeostasis. ITPK isoforms include ITPKA, ITPKB, and ITPKC, all of which contain a conserved catalytic unit in their C termini, but have unique N-terminal sequences and tissue distributions.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Inositol 1,4,5-trisphosphate 3-kinase C; Inositol-trisphosphate 3-kinase C; InsP 3 kinase C; insP 3-kinase C; IP3 3-kinase C; IP3K C
Gene Aliases: IP3-3KC; IP3KC; ITPKC
UniProt ID: (Human) Q96DU7
Entrez Gene ID: (Human) 80271
Molecular Function:
kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support