Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 1
Endotoxin level < 0.01EU/µg by LAL method
Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.
Idarucizumab is a humanized monoclonal antibody fragment (Fab) derived from an immunoglobulin G1 isotype molecule that binds to and inactivates the oral anticoagulant dabigatran, thereby reversing its anticoagulant effect. As a direct acting oral anticoagulant (DOAC), one of the risks associated with the use of dabigatran includes bleeding, especially when given to patients at increased risk (elderly, chronic kidney disease, concomitant NSAID or warfarin use, etc.). Approved under the tradename Praxbind (FDA), idarucizumab is indicated for the emergency treatment of dabigatran-associated bleeding in life-threatening or surgically induced situations. Its use is associated with immediate, complete and sustained reversal of the anticoagulant effects of dabigatran. Idarucizumab protein structure can be viewed below, with disulfide bridges at the following points: H22-H95, H149-H205, H225-L-219, L23-L93, L139-L199.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: aDabi-Fab; dabigatran; dabigatran etexilate mesylate
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support