Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Immunogen sequence: SLNPDEMRRR REEEGLQLRK QKREEQLFKR RNVATAEEET EEEVMSDGGF HEAQINNMEM APGGVITSDM IEMIFSKSPE QQLSATQKFR KLLSKEPNPP IDEVISTPGV VARFVEFLKR KENCTLQFES AWVLTNIASG NSLQTRIVIQ AGAVPIFIEL LSSEFEDVQE QAVWALGNIA GDSTMCRDYV LDCNILPPLL QLFSKQNRLT MTRNAVWALS NLCRGKSPPP EFAKVSPCLN VLSWLLFVSD TDVLA
Recombination activating proteins RAG1 and RAG2 regulate and mediate VJ recombination, the process by which genes for immunoglobulins and T-cell receptors are generated. Several other ubiquitously expressed proteins are thought to be recruited in the recombination process. Among these are the genes affected in severe combined immune deficiency and genes involved in ds-DNA break repair. The protein encoded by this gene interacts with RAG1 and may play a role in VJ recombination. Two transcript variants, one protein-coding and the other not, have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: IMA2; Importin alpha 2; importin alpha 5; importin alpha-5; Importin alpha-S1; importin subunit alpha-1; Importin subunit alpha-5; importin-alpha-S1; IPOA 1; IPOA1; karyopherin (importin) alpha 1; Karyopherin A2; karyopherin alpha 1 (importin alpha 5); Karyopherin subunit alpha-1; karyopherin/importin alpha-1; KPNA2; m-importin-alpha-S1; NPI-1; Nucleoprotein interactor 1; RAG cohort 1; RAG cohort protein 2; RCH 1; RCH1; recombination activating gene cohort 2; SRP 1; SRP1 alpha; SRP1-beta
Gene Aliases: AW494490; IPOA5; KPNA1; mSRP1; NPI-1; NPI1; RCH2; SRP1
UniProt ID: (Human) P52294, (Mouse) Q60960
Entrez Gene ID: (Human) 3836, (Mouse) 16646, (Rat) 288064
Molecular Function:
transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support