Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
Positive Samples: Raji, U-87MG, MCF7, HeLa, HepG2, mouse testis, mouse thymus, LO2, Mouse testis, Mouse lung, Mouse brain, Rat liver; Cellular Location: Cytoplasm, Nucleus envelope
Immunogen sequence: AAEQGRPPEH TSKFYAKGAL QYLVPILTQT LTKQDENDDD DDWNPCKAAG VCLMLLATCC EDDIVPHVLP FIKEHIKNPD WRYRDAAVMA FGCILEGPEP S
Nucleocytoplasmic transport, a signal- and energy-dependent process, takes place through nuclear pore complexes embedded in the nuclear envelope. The import of proteins containing a nuclear localization signal requires the NLS import receptor, a heterodimer of importin alpha and beta subunits also known as karyopherins. Importin alpha binds the NLS-containing cargo in the cytoplasm and importin beta docks the complex at the cytoplasmic side of the nuclear pore complex. In the presence of nucleoside triphosphates and the small GTP binding protein Ran, the complex moves into the nuclear pore complex and the importin subunits dissociate. Importin alpha enters the nucleoplasm with its passenger protein and importin beta remains at the pore. Interactions between importin beta and the FG repeats of nucleoporins are essential in translocation through the pore complex. The protein encoded by this gene is a member of the importin beta family.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HGNC:6400; importin 1; importin 90; Importin beta; importin beta-1 subunit; Importin subunit beta-1; Importin-90; KapB1; karyopherin (importin) beta 1; karyopherin beta 1; Karyopherin subunit beta-1; karyopherin,beta 1; MGC2155; MGC2156; MGC2157; Nuclear factor p97; Pore targeting complex 97 kDa subunit; PTAC97; SCG
Gene Aliases: AA409963; IMB1; Impnb; IPO1; IPOB; KPNB1; NTF97
UniProt ID: (Human) Q14974, (Rat) P52296, (Mouse) P70168
Entrez Gene ID: (Human) 3837, (Rat) 24917, (Mouse) 16211
Molecular Function:
transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support