Search Thermo Fisher Scientific
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%).
This antibody has specificity for H11N9 (A/mallard/Alberta/294/1977) HA/Hemagglutinin.
Protein Aliases: flu hemagglutinin; influenza hemagglutinin