Invitrogen
FIGURE: 1 / 1
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%).
This antibody has specificity for Influenza A H4N6 (A/mallard/Ohio/657/2002) Neuraminidase/NA.