Invitrogen
This antibody specifically binds the hemagglutinin of avian influenza virus subtype H5.
Haemagglutinin is an antigenic glycoprotein which allows viral attachment and entry into the cell. Sixteen subtypes of haemagglutinin (H1-H16) have been described, of which H1, H2 and H3 infect humans. H5 and H7 normally result in avian disease, with some highly pathogenic H5N1 strains causing 100% mortality in poultry. H5N1 has in some cases mutated to infect humans, with 60% mortality. Haemagglutinin H5 is from the Influenza A virus H5N1. Influenza A belongs to the Orthomyxoviridae family, and is a negative sense single-stranded RNA virus which results in respiratory disease. Haemagglutinin is one of the most medically relevant antigens on influenza as it is a target for antiviral drugs and antibodies.
Protein Aliases: Influenza H5; Influenza NP-A