Invitrogen
FIGURE: 1 / 1
H5.3 binds to H5 at the head domain at an epitope which involves the 190 loop. The epitope was determined to be LGVSSA (aa 134-139) and YQGKSS (aa 142-147) of the head domain by DXMS (deuterium exchange mass spectrometry). This antibody shows reactivity with the wild-type H5N1 field strains VN/1203 (EC50 ~ 17 ng/mL), HK/156 (EC50 ~ 18 ng/mL), Anhui (EC50 ~ 2.8 µg/mL) and Indo (EC50 ~ 10 µg/mL). HA is a viral glycoprotein initially assembled as a trimer on the viral envelope as the preprotein HA0. It is subsequently cleaved by host proteases.
Influenza A virus is a major public health threat, killing more than 30,000 people per year in the USA. In early 2009, a novel swine-origin influenza A (H1N1) virus was identified in specimens obtained from patients in Mexico and the United States. The virus spread quickly around the world and on June 11, 2009, the World Health Organization declared it a pandemic. Influenza A virus has one of sixteen possible Hemagglutinin (HA) surface proteins and one of nine possible Neuraminidase (NA) surface proteins. The Hemagglutinin protein facilitates viral attachment while Neuraminidase is involved in viral release. These proteins also elicit immune responses that prevent infection or independently reduce viral replication. The genetic make-up of this swine flu virus is unlike any other: it is an H1N1 strain that combines a triple assortment first identified in 1998 including human, swine, and avian influenza with two new pig H3N2 virus genes from Eurasia, themselves of recent human origin. The distinct antigenic properties of the new swine influenza virus compared with seasonal influenza A (H1N1) virus suggest that human immunity against new swine influenza virus is limited, although the age distribution of reported cases suggests some degree of protection in older age groups.
Protein Aliases: flu hemagglutinin; influenza hemagglutinin