Invitrogen
This antibody binds hemaglutinin of most group 1 influenza viruses. CR6261 recognizes a highly conserved helical region in the membrane-proximal stem of HA1/HA2.
Infectious diseases are the leading cause of death worldwide. AIDS, tuberculosis (TB), malaria, diarrhoeal and respiratory infections account for 78% of deaths caused by infectious disease. As many infectious diseases are controlled, new diseases emerge and old diseases become resistant to current drugs. Many infectious diseases have been associated with an increase risk of carcinoma. Influenza continues to attract researchers as new strains appear by the ability of the influenza gene to mix with different forms of the virus. Recently, research on SARS and West Nile virus has risen due to the increased number of infections. These antibodies assist in research by detecting the infectious disease agent.
Protein Aliases: flu hemagglutinin; influenza hemagglutinin