Invitrogen
FIGURE: 1 / 1
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%).
This antibody has specificity for Influenza B (B/Massachusetts/03/2010) Hemagglutinin/HA1.
Protein Aliases: flu hemagglutinin; influenza b hemagglutinin; influenza hemagglutinin