Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Antibody detects endogenous levels of total IFN10.
Produced by macrophages, IFN-alpha have antiviral activities. Interferon stimulates the production of two enzymes: a protein kinase and an oligoadenylate synthetase.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: IFN alpha; IFN-alpha-10; Interferon alpha-10; Interferon alpha-6L; Interferon alpha-C; interferon, alpha 10; LeIF C
Gene Aliases: IFN-alphaC; IFNA10
UniProt ID: (Human) P01566
Entrez Gene ID: (Human) 3446
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support